missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SARDH Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563523
This item is not returnable.
View return policy
Description
Immunogen sequence: EETGLHTGWI QNGGLFIASN RQRLDEYKRL MSLGKAYGVE SHVLSPAETK TLYPLMNVDD LYGTLYVPHD GTMDPAGTCT TLARAASARG AQVIENCPVT GIRVWTDDFG VRRVAGVETQ HGSI Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 92%.
The function remains known.
Specifications
| SARDH | |
| Polyclonal | |
| Unconjugated | |
| SARDH | |
| BPR-2; dimethylglycine dehydrogenase-like 1; DMGDHL1; FLJ36475; RGD:621125}; SAR; sarcosine dehydrogenase; sarcosine dehydrogenase, mitochondrial; SARD; Sardh; sardh {ECO:0000312; SDH; similar to sarcosine dehydrogenase; zgc:56363 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1757 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UL12 | |
| SARDH | |
| Recombinant protein corresponding to Human SARDH. Recombinant protein control fragment (Product #RP-92943). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction