missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SARDH Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30570
This item is not returnable.
View return policy
Description
SARDH Polyclonal antibody specifically detects SARDH in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| SARDH | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| BPR-2, dimethylglycine dehydrogenase-like 1, DMGDHL1, FLJ36475, mitochondrial, SAR, sarcosine dehydrogenase, SARD, SDH | |
| between 868 and 918 | |
| 100 μg | |
| Endocrinology, Signal Transduction | |
| 1757 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction