missing translation for 'onlineSavingsMsg'
Learn More

Sarcosine methyl ester, hydrochloride, 97%

Product Code. 10346744 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10346744 50 g Glass bottle 50g
10306464 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10346744 Supplier Thermo Scientific Acros Supplier No. 397500500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13515-93-0 | C4H10ClNO2 | 139.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13515-93-0
Molecular Formula C4H10ClNO2
Molecular Weight (g/mol) 139.58
MDL Number MFCD00038876
InChI Key HQZMRJBVCVYVQA-UHFFFAOYSA-N
Synonym sarcosine methyl ester hydrochloride, methyl 2-methylamino acetate hydrochloride, h-sar-ome.hcl, glycine, n-methyl-, methyl ester, hydrochloride, unii-ypw7yyp04f, ypw7yyp04f, sarcosine methyl ester hcl, methyl n-methylaminoacetate hydrochloride, glycine, n-methyl-, methyl ester, hydrochloride 1:1, sar-ome hcl
PubChem CID 83544
IUPAC Name methyl 2-(methylamino)acetate;hydrochloride
SMILES [H+].[Cl-].CNCC(=O)OC

Specifications

Melting Point 110.0°C to 125.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 50 g
Formula Weight 139.58
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Sarcosine methyl ester, hydrochloride
compliance-icons
Product Identifier
  • Sarcosine methyl ester, hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.