missing translation for 'onlineSavingsMsg'
Learn More

Sarcosine ethyl ester hydrochloride, 98+%

Product Code. 11444454
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444454 25 g 25g
11454454 100 g 100g
11464454 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11444454 Supplier Thermo Scientific Alfa Aesar Supplier No. A13112.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 52605-49-9 | C5H12ClNO2 | 153.61 g/mol

Sarcosine ethyl ester hydrochloride, It is used as intermediate in the synthesis of antienzyme agents for toothpaste. Found to be a marker for prostate cancer bioagression. It can be used to produce N-methyl-N-nitroso-glycine ethyl ester. This reaction will need reagent NaNO2, and the menstruum H2O and aq. acetic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sarcosine ethyl ester hydrochloride, It is used as intermediate in the synthesis of antienzyme agents for toothpaste. Found to be a marker for prostate cancer bioagression. It can be used to produce N-methyl-N-nitroso-glycine ethyl ester. This reaction will need reagent NaNO2, and the menstruum H2O and aq. acetic acid.

Solubility
Soluble in water 0.1 g/mL.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52605-49-9
Molecular Formula C5H12ClNO2
Molecular Weight (g/mol) 153.61
MDL Number MFCD00012506
InChI Key NIDZUMSLERGAON-UHFFFAOYSA-N
Synonym sarcosine ethyl ester hydrochloride, ethyl sarcosinate hydrochloride, ethyl 2-methylamino acetate hydrochloride, h-sar-oet.hcl, ethyl n-methylaminoacetate hydrochloride, me-gly-oet.hcl, n-methylglycine ethyl ester hydrochloride, glycine, n-methyl-, ethyl ester, hydrochloride, ethyl n-methylglycinate hydrochloride, n-methyl glycine ethyl ester hydrochloride
PubChem CID 171173
SMILES [H+].[Cl-].CCOC(=O)CNC

Specifications

Melting Point 122°C to 126°C
Linear Formula CH3NHCH2CO2CH2CH3·HCl
Quantity 25 g
Beilstein 3911182
Sensitivity Hygroscopic
Solubility Information Soluble in water 0.1g/mL.
IUPAC Name hydrogen ethyl 2-(methylamino)acetate chloride
Formula Weight 153.61
Percent Purity ≥98%
Chemical Name or Material Sarcosine ethyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.