missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sarcosine ethyl ester hydrochloride, 97%

Product Code. 10147563 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10147563 50 g Glass bottle 50g
10053660 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10147563 Supplier Thermo Scientific Chemicals Supplier No. 305590500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 52605-49-9 | C5H12ClNO2 | 153.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52605-49-9
Molecular Formula C5H12ClNO2
Molecular Weight (g/mol) 153.61
MDL Number MFCD00012506
InChI Key NIDZUMSLERGAON-UHFFFAOYSA-N
Synonym sarcosine ethyl ester hydrochloride, ethyl sarcosinate hydrochloride, ethyl 2-methylamino acetate hydrochloride, h-sar-oet.hcl, ethyl n-methylaminoacetate hydrochloride, me-gly-oet.hcl, n-methylglycine ethyl ester hydrochloride, glycine, n-methyl-, ethyl ester, hydrochloride, ethyl n-methylglycinate hydrochloride, n-methyl glycine ethyl ester hydrochloride
PubChem CID 171173
SMILES [H+].[Cl-].CCOC(=O)CNC

Specifications

Melting Point 123°C to 129°C
Color White
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Linear Formula CH3NHCH2CO2C2H5·HCl
Quantity 50 g
IUPAC Name hydrogen ethyl 2-(methylamino)acetate chloride
Formula Weight 153.61
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Sarcosine ethyl ester hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.