missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ Sarcosine anhydride, 99%
Specifications
Specifications
| CAS | 5076-82-4 |
| Melting Point | 142°C to 147°C |
| Molecular Formula | C6H10N2O2 |
| MDL Number | MFCD00006519 |
| Quantity | 5 g |
| Beilstein | 123513 |
| Synonym | sarcosine anhydride, sarcosine diketopiperazine, 1,4-dimethyl-2,5-dioxopiperazine, n,n'-dimethyldiketopiperazine, 2,5-piperazinedione, 1,4-dimethyl, sarcosine anhydride, bimol. cyclic, 1,4-dimethyl-2,5-diketopiperazine, 2,5-piperazinedione, 1,4-dimethyl-8ci 9ci, cyclo sar-sar, 1,4-dimethyl-1,4-diazaperhydroine-2,5-dione |
| InChI Key | PKDGRAULLDDTRN-UHFFFAOYSA-N |
| SMILES | CN1CC(=O)N(C)CC1=O |
| IUPAC Name | 1,4-dimethylpiperazine-2,5-dione |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?