missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sarcosine, 98%

Product Code. 11905341 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 11905341

Brand: Thermo Scientific Chemicals 157740025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 107-97-1 | C3H7NO2 | 89.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-97-1
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00004279
InChI Key FSYKKLYZXJSNPZ-UHFFFAOYSA-N
Synonym N-Methylglycine
PubChem CID 1088
ChEBI CHEBI:15611
IUPAC Name 2-(methylamino)acetic acid
SMILES CNCC(O)=O

Specifications

Color Colorless or White to Yellow
Physical Form Crystals or Powder
Quantity 2.5 kg
Infrared Spectrum Authentic
Packaging Plastic bottle
Assay Percent Range 98%
Linear Formula CH3NHCH2CO2H
Water 2% max.
Formula Weight 89.09
Percent Purity ≥97.5% (total base)
Chemical Name or Material Sarcosine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.