missing translation for 'onlineSavingsMsg'
Learn More

Sarcalumenin Antibody (XIIC4) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NB120-2874-0.025ml

 View more versions of this product

Product Code. 18588930

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Sarcalumenin Monoclonal specifically detects Sarcalumenin in Mouse, Canine, Guinea Pig, Rabbit samples. It is validated for Western Blot, Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sarcalumenin
Monoclonal
1 mg/mL
Western Blot 1:500, Immunofluorescence 1:500
SRL
Purified rabbit sarcoplasmic reticulum membrane vesicles.
0.025 mL
Apoptosis, Cancer, Tumor Suppressors
6345
Mouse
Ascites
Western Blot
XIIC4
Unconjugated
sarcalumenin
Mouse
Protein A purified
RUO
Primary
This specifically detects sarcalumenin and the 53 kDa glycoprotein splice variant.
Store at -20°C. Avoid freeze/thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only