missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress [Sar9,Met(O2)11]-Substance P

Product Code. 16461998
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16461998 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16461998 Supplier MedChemExpress Supplier No. HYP1012

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress [Sar9,Met(O2)11]-Substance P is a tachykinin NK1 receptor selective agonist.

Specifications

Purity 99.91%
CAS 110880-55-2
Content And Storage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
For Use With (Application) Neuroscience-Neuromodulation
Molecular Weight (g/mol) 1393.66 g/mol
Product Type Peptides
Quantity 5 mg
Shipping Condition Room temperature
Solubility H2O : 50 mg/mL (35.88 mM∣Need ultrasonic)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.