missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAR405838, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
SAR405838 (MI-77301), an analog of MI-773, is a highly potent and selective MDM2-p53 interaction inhibitor. SAR405838 binds to MDM2 with a Ki of 0.88 nM. SAR405838 induces apoptosis and has potent antitumor activity.
Specifications
Specifications
| Chemical Name or Material | SAR405838 |
| CAS | 1303607-60-4 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C29H34Cl2FN3O3 |
| Quantity | 5 mg |
| Synonym | MI-77301 |
| Solubility Information | DMSO : 100 mg/mL (177.78 mM; Need ultrasonic) |
| SMILES | O=C([C@H](N[C@H]1CC(C)(C)C)[C@H](C2=CC=CC(Cl)=C2F)[C@@]31C(NC4=C3C=CC(Cl)=C4)=O)N[C@@H]5CC[C@@H](O)CC5 |
| Molecular Weight (g/mol) | 562.5 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction