missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAR1B, Mouse anti-Human, Clone: AT1C7, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB16349.50uL
This item is not returnable.
View return policy
Specifications
| SAR1B | |
| Monoclonal | |
| Unconjugated | |
| In PBS, pH 7.4 (10% glycerol, 0.02% sodium azide). | |
| CMRD/GTBPB/SARA2 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| AT1C7 | |
| ELISA Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| SAR1 homolog B (S. cerevisiae) | |
| SAR1B | |
| Recombinant protein corresponding to amino acids 1-198 of human SAR1B from E. coli. | |
| RUO | |
| 51128 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction