missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAR1A Polyclonal Antibody

SAR1A Polyclonal Antibody
Brand: Thermo Scientific PA552362
This item is not returnable.
View return policy
Description
SAR1A Polyclonal Antibody for Western Blot, IHC
Specifications
| SAR1A | |
| Polyclonal | |
| Unconjugated | |
| SAR1A | |
| 1600019H17Rik; COPII-associated small GTPase; GTP-binding protein SAR1a; masra2; SAR1; SAR1 gene homolog A; SAR1 gene homolog A (S. cerevisiae); SAR1 homolog A; SAR1A; SAR1a gene homolog 1; Sara; Sara1; secretion associated Ras related GTPase 1A; secretion associated, Ras related GTPase 1A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56681 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NR31 | |
| SAR1A | |
| Recombinant protein corresponding to Human SAR1A. Recombinant protein control fragment (Product #RP-89507). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction