missing translation for 'onlineSavingsMsg'
Learn More

SAR131675, TRC

Product Code. 30427305
Change view
Click to view available options
Quantity:
10 mg
50 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30427305 10 mg
30413918 500 mg
30420765 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30427305 Supplier Toronto Research Chemicals Supplier No. TRCS13951010MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1092538-80-1
Molecular Formula C18H22N4O4
Molecular Weight (g/mol) 358.39
Synonym 2-Amino-1-ethyl-7-(3-hydroxy-4-methoxy-3-methyl-1-butyn-1-yl)-N-methyl-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxamide, 2-Amino-1-ethyl-1,4-dihydro-7-(3-hydroxy-4-methoxy-3-methyl-1-butyn-1-yl)-N-methyl-4-oxo-1,8-naphthyridine-3-carboxamide
IUPAC Name 2-amino-1-ethyl-7-(3-hydroxy-4-methoxy-3-methylbut-1-ynyl)-N-methyl-4-oxo-1,8-naphthyridine-3-carboxamide
SMILES CCN1C(=C(C(=O)C2=C1N=C(C=C2)C#CC(C)(COC)O)C(=O)NC)N

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C18H22N4O4/c1-5-22-15(19)13(17(24)20-3)14(23)12-7-6-11(21-16(12)22)8-9-18(2,25)10-26-4/h6-7,25H,5,10,19H2,1-4H3,(H,20,24)
Formula Weight 358.1641
Chemical Name or Material SAR131675
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.