missing translation for 'onlineSavingsMsg'
Learn More

SAR125844, MedChemExpress

Product Code. p-200101577
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16473097

Brand: Medchem Express HY16446

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SAR125844 is a potent, highly selective, reversible and ATP-competitive MET receptor tyrosine kinase (RTK) inhibitor, with an IC50 of 4.2 nM. Shows inhibition of MET autophosphorylation in cell-based assays.

SAR125844 is a potent, highly selective, reversible and ATP-competitive MET receptor tyrosine kinase (RTK) inhibitor, with an IC50 of 4.2 nM. Shows inhibition of MET autophosphorylation in cell-based assays.

Specifications

Chemical Name or Material SAR125844
CAS 1116743-46-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C25H23FN8O2S2
Quantity 50 mg
Solubility Information DMSO : 45 mg/mL (81.72 mM; Need ultrasonic)
SMILES O=C(NCCN1CCOCC1)NC2=NC3=CC=C(SC4=NN=C5C=CC(C6=CC=C(F)C=C6)=NN54)C=C3S2
Molecular Weight (g/mol) 550.63
Formula Weight 550.63
Percent Purity 98.11%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.