missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAR125844, MedChemExpress
Description
SAR125844 is a potent, highly selective, reversible and ATP-competitive MET receptor tyrosine kinase (RTK) inhibitor, with an IC50 of 4.2 nM. Shows inhibition of MET autophosphorylation in cell-based assays.
Specifications
Specifications
| Chemical Name or Material | SAR125844 |
| CAS | 1116743-46-4 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C25H23FN8O2S2 |
| Quantity | 50 mg |
| Solubility Information | DMSO : 45 mg/mL (81.72 mM; Need ultrasonic) |
| SMILES | O=C(NCCN1CCOCC1)NC2=NC3=CC=C(SC4=NN=C5C=CC(C6=CC=C(F)C=C6)=NN54)C=C3S2 |
| Molecular Weight (g/mol) | 550.63 |
| Formula Weight | 550.63 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?