missing translation for 'onlineSavingsMsg'
Learn More

SAR1 Antibody, Novus Biologicals™

Product Code. 18336142 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18336142 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18336142 Supplier Novus Biologicals Supplier No. NBP312418

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SAR1 Polyclonal antibody specifically detects SAR1 in Drosophila samples. It is validated for Western Blot, ELISA, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAR1
Applications Western Blot, ELISA, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:10000, Immunoprecipitation 1:200
Formulation Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide
Gene Alias COPII-associated small GTPase, EC 3.6.5, masra2, SAR1 gene homolog A, SAR1 homolog A (S. cerevisiae), SAR1a gene homolog (S. cerevisiae) 1, SAR1a gene homolog 1, SAR1a gene homolog 1 (S. cerevisiae), SAR1SARA1GTP-binding protein SAR1a, Sara
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 320-370 on SARA protein.
Purification Method Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Membrane Trafficking and Chaperones, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 56681
Target Species Drosophila
Content And Storage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.