missing translation for 'onlineSavingsMsg'
Learn More

SAR1 Antibody, Novus Biologicals™

Product Code. 18250641 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18250641 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18250641 Supplier Novus Biologicals Supplier No. NBP102306

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

SAR1 Polyclonal specifically detects SAR1 in Mouse samples. It is validated for Western Blot, Peptide ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAR1
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.1 - 0.3 ug/ml, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_064535.1
Gene Alias COPII-associated small GTPase, EC 3.6.5, masra2, SAR1 gene homolog A, SAR1 homolog A (S. cerevisiae), SAR1a gene homolog (S. cerevisiae) 1, SAR1a gene homolog 1, SAR1a gene homolog 1 (S. cerevisiae), SAR1SARA1GTP-binding protein SAR1a, Sara
Gene Symbols SAR1A
Host Species Goat
Immunogen Peptide with sequence C-HSRLVESKVELNA corresponding to internal region according to NP_064535.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Membrane Trafficking and Chaperones, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 56681
Test Specificity Cross reacts with Mouse. Expected from sequence similarity: Human and Rat.
Target Species Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.