missing translation for 'onlineSavingsMsg'
Learn More

SAR-260301, MedChemExpress

Product Code. 16489397
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16489397 5 mg 5mg
16486858 10 mg 10mg
16466956 100 mg 100mg
16463496 25 mg 25mg
16462927 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16489397 Supplier Medchem Express Supplier No. HY15837

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SAR-260301 is an orally available and selective PI3Kβ inhibitor with an IC50 of 23 nM.

SAR-260301 is an orally available and selective PI3Kβ inhibitor with an IC50 of 23 nM.

Specifications

Chemical Name or Material SAR-260301
Physical Form Solid
CAS 1260612-13-2
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C19H22N4O3
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 125 mg/mL (352.71 mM; Need ultrasonic)
SMILES O=C1N=C(CC(N2[C@@H](C)CC3=C2C=CC=C3)=O)NC(N4CCOCC4)=C1
Molecular Weight (g/mol) 354.4
Formula Weight 354.4
Percent Purity 99.37%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.