missing translation for 'onlineSavingsMsg'
Learn More

SAR-260301, MedChemExpress

Product Code. 16486858
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16486858

Brand: Medchem Express HY15837

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SAR-260301 is an orally available and selective PI3Kβ inhibitor with an IC50 of 23 nM.

SAR-260301 is an orally available and selective PI3Kβ inhibitor with an IC50 of 23 nM.

Specifications

Chemical Name or Material SAR-260301
Physical Form Solid
CAS 1260612-13-2
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C19H22N4O3
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 125 mg/mL (352.71 mM; Need ultrasonic)
SMILES O=C1N=C(CC(N2[C@@H](C)CC3=C2C=CC=C3)=O)NC(N4CCOCC4)=C1
Molecular Weight (g/mol) 354.4
Formula Weight 354.4
Percent Purity 99.37%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.