missing translation for 'onlineSavingsMsg'
Learn More

GDC-0339, MedChemExpress

Product Code. 16409767
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16409767

Brand: Medchem Express HY16976

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GDC-0339 is a potent, orally bioavailable and well tolerated pan-Pim kinase inhibitor, with Kis of 0.03 nM, 0.1 nM and 0.02 nM for Pim1, Pim2 and Pim3, respectively. GDC-0339 is discovered as a potential treatment of multiple myeloma.

GDC-0339 is a potent, orally bioavailable and well tolerated pan-Pim kinase inhibitor, with Kis of 0.03 nM, 0.1 nM and 0.02 nM for Pim1, Pim2 and Pim3, respectively. GDC-0339 is discovered as a potential treatment of multiple myeloma.

Specifications

Chemical Name or Material GDC-0339
CAS 1428569-85-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H22F3N7OS
Quantity 5 mg
Solubility Information DMSO : ≥ 52 mg/mL (111.71 mM)
SMILES FC1=CC=CC(F)=C1C2=NC(C(NC3=C(N4CC[C@@H](F)[C@H](N)CC4)N(C)N=C3)=O)=C(N)S2
Molecular Weight (g/mol) 465.5
Formula Weight 465.5
Percent Purity 99.07%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.