missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Sapphire PCR Tubes with Attached Frosted Flat Cap

Product Code. 13045063
Change view
Click to view available options
Color:
Assorted
Blue
Green
Natural
Red
Violet
Yellow
Volume (Metric):
0.2 mL
0.5 mL
Unit Size:
Pack of 1000
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Volume (Metric) unitSize
13045063 Violet 0.2 mL -
13025053 Blue 0.5 mL -
13035053 Green 0.5 mL Pack of 1000
13045053 Yellow 0.5 mL -
13005063 Red 0.2 mL -
13055053 Violet 0.5 mL -
13015063 Blue 0.2 mL Pack of 1000
13025063 Green 0.2 mL -
13035063 Yellow 0.2 mL Pack of 1000
10507081 Natural 0.5 mL Pack of 1000
13015053 Red 0.5 mL -
13085053 Assorted 0.2 mL Pack of 1000
10247111 Natural 0.2 mL Pack of 1000
13065053 Assorted 0.5 mL Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
Product Code. 13045063 Supplier Greiner Bio-One Supplier No. 683277

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For use in RT/Quantitative-PCR. Greiner Bio-One Sapphire PCR Tubes with Attached Frosted Flat Cap are comprehensive thermal cycler compatible.

  • Thin-walled for heat transfer during amplification
  • Free of detectable DNase, RNase, human DNA, endotoxin and PCR inhibitors
  • Packaging labelled with product symbols for warehousing
  • Used for sample handling
TRUSTED_SUSTAINABILITY

Specifications

Color Violet
Volume (Metric) 0.2 mL
For Use With (Equipment) PCR
Sterility Non-sterile
Material Polypropylene
Closure Type Attached Frosted Flat Cap
Product Type PCR Tubes with Attached Flat Caps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.