missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Sapphire™ Luminescence Enhancer for Solution Assays
Click to view available options
Quantity:
2500 Assays
Unit Size:
Each
Description
For use with chemiluminescent substrates.
Specifications
Specifications
| For Use With (Application) | Chemiluminescence enhancer |
| Content And Storage | Store at 2° to 8°C |
| Product Type | Sapphire Luminescence Enhancer for Solution Assays |
| Quantity | 2500 Assays |
| Format | 96-well plate |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction