missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SAPK4 Monoclonal Antibody (R.618.4)

Rabbit Monoclonal Antibody
Brand: Invitrogen™ MA514994
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with other isoforms of p38 MAPK.
MAPK13 (p38 delta) is a serine/threonine kinase that transduces a variety of extracellular signals, including cytokine and environmental stress responses. MAPK13 also regulates the differentiation of keratinocytes. This kinase is activated by various environmental stresses and proinflammatory cytokines. The activation requires its phosphorylation by MAP kinase kinases, or its autophosphorylation triggered by the interaction of MAP3K7IP1/TAB1 protein with this kinase. The substrates of this kinase include transcription regulator ATF2, MEF2C, and MAX, cell cycle regulator CDC25B, and tumor suppressor p53, which suggest the roles of this kinase in stress related transcription and cell cycle regulation, as well as in genotoxic stress response. Four alternatively spliced transcript variants of SAPK4 encoding distinct isoforms have been reported.
Specifications
| SAPK4 | |
| Monoclonal | |
| Unconjugated | |
| MAPK13 | |
| im:7136778; MAP kinase 13; MAP kinase p38 delta; MAPK 13; Mapk13; MAPK-13; mitogen activated protein kinase 13; mitogen-activated protein kinase 13; Mitogen-activated protein kinase p38 delta; p38 delta MAP kinase; p38delta; PRKM13; SAPK/Erk/kinase 4; SAPK4; Serk4; Stress-activated protein kinase 4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 29513, 5603 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| R.618.4 | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5 | |
| O15264, Q9WTY9 | |
| MAPK13 | |
| Synthetic peptide corresponding to residues around the carboxy terminus of p38d MAPK. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction