missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SAPAP3 Polyclonal Antibody

Product Code. 12858642
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12858642 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12858642 Supplier Invitrogen™ Supplier No. PA520465

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is rat brain tissue lysate. PA5-20465 can be used with blocking peptide PEP-0582.

SAP90/PSD-95-associated protein 3 (SAPAP3, also known as DLGAP3) is a member of a protein family whose members specifically interact with PSD-95/SAP90, a membrane-associated guanylate kinase localized at postsynaptic density (PSD) in neuronal cells. Like the other SAPAP proteins, SAPAP3 is thought to be an adaptor protein that also interacts with different synaptic scaffolding proteins, cytoskeletal and signaling components, such as focal adhesion kinase (FAK) and proline-rich tyrosine kinase 2 (PYK2). Both SAPAP3 protein and mRNA are targeted to dendrites, whereas SAPAP1, -2, and -4 mRNAs are detected mainly in cell bodies. Recent experiments have suggested that SAPAP3 may be involved in obsessive-compulsive disorder (OCD), as mice lacking SAPAP3 exhibited OCD-like symptoms which could be relieved by lentiviral-mediated selective expression of SAPAP3 in the striatum of SAPAP3-mutant mice. At least two isoforms are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAPAP3
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene DLGAP3
Gene Accession No. O95886, P97838, Q6PFD5
Gene Alias BC058433; DAP3; DAP-3; discs large homolog associated protein 3; discs, large (Drosophila) homolog-associated protein 3; disks large-associated protein 3; DLG associated protein 3; Dlgap3; Prpl8; PSD-95/SAP90-associated protein-3; PSD-95/SAP90-binding protein 3; SAP90/PSD 95 associated protein 3; SAP90/PSD-95-associated protein 3; SAPAP3
Gene Symbols DLGAP3
Host Species Rabbit
Immunogen An 18 amino acid peptide from near the carboxy terminus of human SAPAP3.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 242667, 286923, 58512
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.