missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SAPAP1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12808712
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12808712 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12808712 Supplier Invitrogen™ Supplier No. PA520531

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is human brain tissue lysate. PA5-20531 can be used with blocking peptide PEP-0651.

Part of the postsynaptic scaffold in neuronal cells. Target binds the guanylate kinase-like domain of the PSD-95 family of synaptic ion channel clustering proteins; may play a role in protein-protein interactions at the synaptic junction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAPAP1
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Dlgap1
Gene Accession No. O14490, P97836, Q9D415
Gene Alias 4933422O14Rik; 9630002F18; AI845682; AI848168; BB075781; D17Bwg0511e; DAP1; DAP-1; DAP-1 beta; DAP-1-ALPHA; DAP-1-BETA; discs large homolog associated protein 1; discs, large (Drosophila) homolog-associated protein 1; disks large-associated protein 1; DLG associated protein 1; Dlgap1; DLGAP1A; DLGAP1B; GKAP; GKAP/SAPAP; GKAP/SAPAP1; GKPA/SAPAP; guanylate kinase associated protein; guanylate kinase-associated protein; hGKAP; Kiaa4162; mKIAA4162; PSD-95 binding protein; PSD-95/SAP90 binding protein 1; PSD-95/SAP90-binding protein 1; rGKAP; SAP90/PSD-95 associated protein 1; SAP90/PSD-95-associated protein 1; SAPAP1
Gene Symbols Dlgap1
Host Species Rabbit
Immunogen An 18 amino acid peptide from near the center of human SAPAP1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 224997, 65040, 9229
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.