missing translation for 'onlineSavingsMsg'
Learn More

SAP97 Polyclonal Antibody
GREENER_CHOICE

Product Code. 11386763
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11386763 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11386763 Supplier Invitrogen Supplier No. PA518790
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

SAP97 Polyclonal Antibody

SAP97 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen SAP97
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene DLG1
Gene Accession No. Q12959
Gene Alias B130052P05Rik; discs large homolog 1; discs large homolog 1, scribble cell polarity complex component; discs large MAGUK scaffold protein 1; discs, large homolog 1; discs, large homolog 1 (Drosophila); discs, large homolog 1 (Drosophila) transcript variant 6-v1; discs, large homolog 1 (Drosophila) transcript variant 6-v2; disks large homolog 1; dJ1061C18.1.1; DKFZp761P0818; DKFZp781B0426; DLG1; Dlgh1; Drosophila discs-large tumor suppressor homologue (synapse associated protein); E-dlg/SAP97; Embryo-dlg/synapse-associated protein 97; hDlg; mKIAA4187; presynaptic protein SAP97; SAP97; SAP-97; synapse-associated protein 97
Gene Symbols DLG1
Host Species Goat
Immunogen Synthetic peptide sequence (NSKTRDKGEIPDD) corresponding to the internal amino acids of DLG1 (aa 661-674).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1739
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.