missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAP30 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99031-50ul
This item is not returnable.
View return policy
Description
SAP30 Polyclonal antibody specifically detects SAP30 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| SAP30 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 | |
| 30 kDa Sin3-associated polypeptide, histone deacetylase complex subunit SAP30, Sin3 corepressor complex subunit SAP30, Sin3A-associated protein, 30kDa, Sin3-associated polypeptide p30, sin3-associated polypeptide, 30 kDa, Sin3-associated polypeptide, 30kDa | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human SAP30. | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 8819 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction