missing translation for 'onlineSavingsMsg'
Learn More

SAP30 Antibody, DyLight 755, Novus Biologicals™

Product Code. 18431546 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18431546 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18431546 Supplier Novus Biologicals Supplier No. NBP192648IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SAP30 Polyclonal antibody specifically detects SAP30 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAP30
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 755
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias 30 kDa Sin3-associated polypeptide, histone deacetylase complex subunit SAP30, Sin3 corepressor complex subunit SAP30, Sin3A-associated protein, 30kDa, Sin3-associated polypeptide p30, sin3-associated polypeptide, 30 kDa, Sin3-associated polypeptide, 30kDa
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 170 and 220 of human Sin3A-Associated Protein, 30kDa using the numbering given in entry NP_003855.1 (GeneID 8819).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8819
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.