missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAP155 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-55255
This item is not returnable.
View return policy
Description
SAP155 Polyclonal specifically detects SAP155 in Human samples. It is validated for Western Blot, Simple Western, Immunoprecipitation, Knockdown Validated.
Specifications
| SAP155 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| Hsh155, pre-mRNA processing 10, pre-mRNA splicing factor SF3b, 155 kDa subunit, Pre-mRNA-splicing factor SF3b 155 kDa subunit, Prp10, PRPF10, SAP 155, SAP155splicing factor 3b, subunit 1, 155kD, SF3B155, SF3b155PRP10, spliceosome associated protein 155, Spliceosome-associated protein 155, splicing factor 3B subunit 1, splicing factor 3b, subunit 1, 155kDa | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 23451 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation, KnockDown | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Simple Western, Immunoprecipitation 2-5 ug/mg lysate, Knockdown Validated | |
| O75533 | |
| SF3B1 | |
| The immunogen recognized by this antibody maps to a region between residue 100 and 150 of human splicing factor 3b, subunit 1, 155kDa using the numbering given in entry NP_036565.2 (GeneID 23451). | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction