missing translation for 'onlineSavingsMsg'
Learn More

SAP130 Rabbit anti-Human, Mouse, Rat, Clone: 3K1X4, Novus Biologicals™

Product Code. 18385623 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18385623 20 μg 20µL
18311084 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18385623 Supplier Novus Biologicals Supplier No. NBP31684220UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

SAP130 Monoclonal antibody specifically detects SAP130 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAP130
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3K1X4
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias KIAA0017splicing factor 3b, subunit 3, 130kD, Pre-mRNA-splicing factor SF3b 130 kDa subunit, RSE1, SAP130SAP 130, SF3B130, SF3b130pre-mRNA splicing factor SF3b, 130 kDa subunit, Spliceosome-associated protein 130, splicing factor 3B subunit 3, splicing factor 3b, subunit 3, 130kDa, STAF130
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1101-1217 of human SAP130 (Q15393).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23450
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.