missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SAP130 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521772
This item is not returnable.
View return policy
Description
Recommended positive controls: HeLa. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SAP130 is a subunit of the histone deacetylase-dependent SIN3A corepressor complex (Fleischer et al., 2003).
Specifications
| SAP130 | |
| Polyclonal | |
| Unconjugated | |
| Sap130 | |
| 130 kDa Sin3-associated polypeptide; 2610304F09Rik; 6720406D06; FLJ12761; Histone deacetylase complex subunit SAP130; KIAA0017; mSin3A-associated protein 130; pre-mRNA splicing factor SF3b, 130 kDa subunit; pre-mRNA-splicing factor SF3b 130 kDa subunit; RGD1311657; RSE1; SAP 130; SAP130; SF3b130; SF3B3; Sin3A associated protein; Sin3A associated protein 130; Sin3A associated protein 130kDa; Sin3A associated protein, 130kDa; Sin3A-associated protein, 130kDa; Sin3-associated polypeptide p130; spliceosome-associated protein 130; Splicing factor 3B subunit 3; STAF130 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79595 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9H0E3 | |
| Sap130 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human SAP130. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction