missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAP130 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31281
This item is not returnable.
View return policy
Description
SAP130 Polyclonal specifically detects SAP130 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| SAP130 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| SF3B3 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human SAP130. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 23450 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0mg/mL | |
| Western Blot 1:500-1:20000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| KIAA0017splicing factor 3b, subunit 3, 130kD, Pre-mRNA-splicing factor SF3b 130 kDa subunit, RSE1, SAP130SAP 130, SF3B130, SF3b130pre-mRNA splicing factor SF3b, 130 kDa subunit, Spliceosome-associated protein 130, splicing factor 3B subunit 3, splicing factor 3b, subunit 3, 130kDa, STAF130 | |
| Rabbit | |
| 136 kDa | |
| 100 μL | |
| Primary | |
| Tested in human. Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction