missing translation for 'onlineSavingsMsg'
Learn More

SAP102 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NB100-2063-0.025mg

 View more versions of this product

Product Code. 18527780

  • £159.00 / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SAP102 Polyclonal specifically detects SAP102 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SAP102
Polyclonal
Unconjugated
PBS with 1 mg/ml BSA with 0.05% Sodium Azide
DLG3
Synthetic peptide corresponding to residues G(83) P V P P K P V P G K N T P K L N G(100) of rat SAP102.
0.025 mg
Cell Cycle and Replication, Neuroscience
1741
Mouse
IgG
Western Blot
1 mg/mL
Western Blot 3 μg/mL, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Frozen 10 μg/mL
discs, large homolog 3 (Drosophila), KIAA1232discs, large homolog 3 (neuroendocrine-dlg, Drosophila), MRX90MRX, NEDLGdisks large homolog 3, neuroendocrine-DLG, SAP-102, SAP102NE-Dlg, Synapse-associated protein 102, XLMR
Rabbit
Affinity Purified
RUO
Primary
SAP102
Store at -20°C. Avoid freeze/thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only