missing translation for 'onlineSavingsMsg'
Learn More

SAP102 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£222.00 - £470.00

Specifications

Antigen SAP102
Dilution Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18412242
View Documents
Novus Biologicals
NBP1-87691
0.1 mL
£470.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18402812
View Documents
Novus Biologicals
NBP1-87691-25ul
25 μL
£222.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SAP102 Polyclonal antibody specifically detects SAP102 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

SAP102
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cell Cycle and Replication, Neuroscience
PBS (pH 7.2) and 40% Glycerol
1741
IgG
Immunogen affinity purified
Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Polyclonal
Purified
RUO
Human
discs, large homolog 3 (Drosophila), KIAA1232discs, large homolog 3 (neuroendocrine-dlg, Drosophila), MRX90MRX, NEDLGdisks large homolog 3, neuroendocrine-DLG, SAP-102, SAP102NE-Dlg, Synapse-associated protein 102, XLMR
This antibody was developed against Recombinant Protein corresponding to amino acids: HKHQHCCKCPECYEVTRLAALRRLEPPGYGDWQVPDPYGPGGGNGASAGYGGYSSQTLPSQAGATPTPRTKAKLIPTGRDVGPVPPKPVPGKSTPKLNGSGPSWWPECTCTNRDWYEQVNGSD
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.