Learn More
SAP 97 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29292-1-AP-20UL
This item is not returnable.
View return policy
Description
The SAP 97 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SAP 97. This antibody recognizes human antigen. The SAP 97 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| SAP 97 | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Human |
| Western Blot, Immunohistochemistry, ELISA | |
| Unconjugated | |
| 20 μL | |
| 1739 | |
| IgG |