missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Santacruzamate A

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Santacruzamate A is a highly potent and selective histone deactylase 2 (HDAC2) inhibitor (IC50 = 112 - 119 pM). Also inhibits HDAC6 in nanomolar range (IC50 = 433 nM). Displays >3500 fold selectivity for HDAC2 over HDAC6, and >8500-fold selectivity over HDAC4 (IC50 >1 μM for HDAC4). Inhibits growth of HuT-78 cutaneous T-cell lymphoma cell line. Identified as targeting human host proteins that interact with SARS-CoV-2.
Specifications
Specifications
| Chemical Name or Material | Ethyl N-[4-oxo-4-[(2-phenylethyl)amino]butyl]carbamate |
| CAS | 1477949-42-0 |
| Quantity | 10 mg |
| Target | Class II Histone Deacetylases Inhibitors |
| Molecular Formula | C15H22N2O3 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction