Learn More
Tocris Bioscience™ SANT-2
Inhibitor of Hedgehog (Hh) signaling; antagonizes smoothened activity
Specifications
Purity >98%
View More Specs
Description
Description
Inhibitor of Sonic hedgehog (Shh) signaling; antagonizes smoothened receptor activity (KD = 12 nM). Displays allosteric binding characteristics similar to SANT-1 (Cat. No. 1974). Displaces smo-[3H]SAG-1.3 and -[3H]Cyclopamine binding (Ki values are 7.8 nM and 8.4 nM respectively).
Specifications
Specifications
>98%