missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Sanitary PVDF Clamps

Product Code. 11719589
Change view
Click to view available options
Size (Metric) Grip:
25.4 to 38.1 mm
50.8 mm
63.5 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size (Metric) Grip unitSize
11719589 50.8 mm Each
11709589 25.4 to 38.1 mm Each
15352830 63.5 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11719589 Supplier Masterflex™ Supplier No. 3181102

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Autoclavable Autoclavable
Quantity 1 Ea.
Type Clamp
Material PVDF
For Use With (Equipment) Sanitary Fitting
Size (Metric) Grip 50.8 mm
Temperature (Metric) Resistant 138°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.