Learn More
Value Plastics Sanitary Fitting Caps, Polypropylene
Shop All Cole-Parmer Products
Click to view available options
Inlet:
1-1/2 in. Sanitary Cap
3/4 in. Mini Sanitary Cap
Unit Size:
Pack of 100
Description
Ideal for capping off fluid transfer system. Sanitary fittings provide a secure connection where preventing contamination is crucial. Constructed of ADCF polypropylene
Specifications
Specifications
| Certifications/Compliance | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Packaging Quantity | 100/Pk. |
| Inlet | 3/4 in. Mini Sanitary Cap |
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.