Learn More
Masterflex™ Sanitary Clamp to Hose Barb Straight Adapter Fittings
Click to view available options
Includes:
1.5 in. Tri-Clamp
Mini Tri-Clamp
Unit Size:
Pack of 6
Description
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Meet requirements for USP Class VI, FDA, REACH, and California Proposition 65
- Surface finishes to 15 micron minimizes adhesion or entrapment
- Packaged to exacting tolerances under ISO Class 7 cleanroom conditions
- Compatible with other Tri-Clamp™ connections
- The sanitary Tri-Clamp barbed adapters are ideal for use in the biopharmaceutical, food and beverage, and single-use disposable industries.
- The fittings are made from animal-derived ingredient-free and phthalate-free polypropylene (PP)
- They have excellent chemical resistance and may be sterilized by gamma irradiation, EtO, E-beam, or autoclave
Specifications
Specifications
| Material | Polypropylene |
| Packaging Quantity | 6 |
| Hose Barb | 1/4 in. |
| Includes | Mini Tri-Clamp |
| Product Type | Sanitary Barbed Adapter |