Learn More
Labplas Sani-Sponge Sampling Bags, Regular tabs: With Clear Body, Tear-Off Protection Strip
Product Code. 17976469
Change view
Click to view available options
Includes:
DE Neutralizing buffer
Unit Size:
Pack of 100
Product Code. Includes unitSize
17976469 DE Neutralizing buffer Pack of 100
1 options
Product Code. 17976469
Brand: Labplas KSS67110DE
Designed to collect samples to detect the presence of microbiological contaminations
  • Detects Listeria, Salmonella and E. coli
  • Contaminant-free
  • Virgin polyethylene
  • Large bag mouth opening
  • Easy to use pull tabs
  • Sponge for swabbing
  • Dehydrated sponge
Specifications
Description DE Neutralizing buffer
Includes DE Neutralizing buffer
Volume (Metric) 450 mL
Material Polyethylene