missing translation for 'onlineSavingsMsg'
Learn More

Sanguinarine chloride, MedChemExpress

Product Code. p-200100087
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16441288

Brand: Medchem Express HYN0052A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sanguinarine (Sanguinarin) chloride, a benzophenanthridine alkaloid derived from the root of Sanguinaria Canadensis, can stimulate apoptosis via activating the production of reactive oxygen species (ROS). Sanguinarine-induced apoptosis is associated with the activation of JNK and NF-κB.

Sanguinarine (Sanguinarin) chloride, a benzophenanthridine alkaloid derived from the root of Sanguinaria Canadensis, can stimulate apoptosis via activating the production of reactive oxygen species (ROS). Sanguinarine-induced apoptosis is associated with the activation of JNK and NF-κB.

Specifications

Chemical Name or Material Sanguinarine chloride
CAS 5578-73-4
Color Red
Purity Grade Notes Research
Molecular Formula C20H14ClNO4
Quantity 5 mg
Synonym Sanguinarin chlorideSanguinarium chloridePseudochelerythrine chloride
Solubility Information H2O : 6 mg/mL (16.31 mM; Need ultrasonic) ∣DMSO : 5 mg/mL (13.60 mM; Need ultrasonic)
SMILES C[N+]1=CC2=C3C(OCO3)=CC=C2C(C=CC4=C5)=C1C4=CC6=C5OCO6.[Cl-]
Molecular Weight (g/mol) 367.78
Formula Weight 367.78
Percent Purity 96.33%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.