missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Sanguinarine chloride

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
Sanguinarine chloride is an inhibitor of protein phosphatase 2C (PP2C), selective over PP1, PP2A and PP2B in vitro. Also inhibits mitogen-activated protein kinase phosphatase-1 (MKP-1) (IC50 = 10 μM). Plant alkaloid that exhibits anti-inflammatory and antioxidant activity. Blocks cell cycle in the G1 phase; inhibits cell growth in human prostate carcinoma cells.
Specifications
Specifications
| Chemical Name or Material | 13-Methyl-[1,3]-benzodioxolo[5,6-c]-1,3-dioxolo[4,5-i]phenanthridinium chloride |
| CAS | 5578-73-4 |
| Assay Percent Range | >97% |
| Quantity | 25 mg |
| Molecular Formula | C20H14ClNO4 |
| Formula Weight | Observed MW: 367.78 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction