missing translation for 'onlineSavingsMsg'
Learn More

Sand, White Quartz, Honeywell Fluka™

Product Code. 15672910 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
50 lb.
Unit Size:
1kg
500g
50lb
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15672910 500 g 500g
15682910 50 lb. 50lb
15662910 1 kg 1kg
15692910 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15672910 Supplier Honeywell-Fluka Supplier No. 274739500G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

50-70 mesh particle size

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14808-60-7
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
SMILES O=[Si]=O

Specifications

Color White
Quantity 500 g
Linear Formula SiO2
UN Number NONH for all modes of transport
IUPAC Name dioxosilane
Formula Weight 60.08g/mol
Chemical Name or Material Sand, White Quartz
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.