missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sand, washed, Thermo Scientific Chemicals
Click to view available options
Quantity:
1000 g
5000 g
Description
Sand, washed is employed in the extraction of DNA and RNA due to its ability to bind to the nucleic acids under the presence of chaotropes. It may be employed as insulating layer on top of the silicon in the fabrication of gold electrode, used for flow injection amperometric detection of organophosphorus (OP) nerve agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sand, washed is employed in the extraction of DNA and RNA due to its ability to bind to the nucleic acids under the presence of chaotropes. It may be employed as insulating layer on top of the silicon in the fabrication of gold electrode, used for flow injection amperometric detection of organophosphorus (OP) nerve agents.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Store in cool, dry conditions in well sealed containers.
Sand, washed is employed in the extraction of DNA and RNA due to its ability to bind to the nucleic acids under the presence of chaotropes. It may be employed as insulating layer on top of the silicon in the fabrication of gold electrode, used for flow injection amperometric detection of organophosphorus (OP) nerve agents.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Store in cool, dry conditions in well sealed containers.
Specifications
Boiling Point | 2230°C |
Melting Point | 1710°C |
Color | Brown-Cream |
Physical Form | Granules |
Quantity | 5000 g |
Linear Formula | SiO2 |
Solubility Information | Insoluble in water. |
Formula Weight | 60.08 |
Odor | Odorless |
Chemical Name or Material | Sand, Washed |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'