Learn More
OHAUS™ Sand Bath
Product Code. 15752589 Shop All Ohaus Products
Click to view available options
No. of Blocks:
1
2
4
6
Unit Size:
Each
Product Code. 15752589
Brand: OHAUS™ 30400173
Only null left
Add to basket
Add to basket
OHAUS™ Sand Bath is constructed of stainless steel for durability.
  • Stainless steel sand baths are available for irregular samples when there is no suitable block available
  • Low temperature covers help to reduce the air flow for additional temperature stability
TRUSTED_SUSTAINABILITY
Specifications
Holds 1 Block
Material Stainless Steel
Type Sand Bath
Includes Thermometer Well
No. of Blocks 1
Size 1 Block