missing translation for 'onlineSavingsMsg'
Learn More

Sancycline, MedChemExpress

Product Code. 16489837
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16489837 5 mg 5mg
16483377 50 mg 50mg
16457358 10 mg 10mg
16423576 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16489837 Supplier Medchem Express Supplier No. HY17466
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Sancycline is a rare semi-synthetic tetracycline prepared by hydrogenolysis of the chloro and benzylic hydroxy moieties of Declomycin.

Sancycline is a rare semi-synthetic tetracycline prepared by hydrogenolysis of the chloro and benzylic hydroxy moieties of Declomycin.

Specifications

Chemical Name or Material Sancycline
CAS 808-26-4
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C21H22N2O7
Quantity 5 mg
Synonym Bonomycin 6-Demethyl-6-deoxytetracycline
Solubility Information DMSO : 8.33 mg/mL (20.10 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(C(C1=O)=C(O)[C@@H](N(C)C)[C@]2([H])C[C@]3([H])CC4=C(C(C3=C(O)[C@@]21O)=O)C(O)=CC=C4)N
Molecular Weight (g/mol) 414.41
Formula Weight 414.41
Percent Purity 99.12%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.