Learn More
VITLAB™ SAN Volumetric Beaker
Measure, dispense and dispose of lab solutions with ease. VITLAB™ SAN Volumetric Beaker is constructed of sturdy, crystal clear styrene acrylonitrile with raised graduations, a convenient pour spout and a large, easy-grip handle. Available in a variety of sizes.
Brand:  VITLAB™ 44191
Product Code. 10030621
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
These measuring beakers feature a convenient spout and large handle to make measuring, pouring and disposing of solutions easy and mess-free. Lightweight and completely transparent acrylic beakers are ideal for a variety of lab purposes.
  • Stable, easy-grip handle and pour spout for secure handling and mess-free dispensing
  • Constructed of strong, shatter-resistant SAN that is resistant to boiling and chemically inert
  • Crystal clear surface with a raised graduated scale for maximum visibility
  • Raised scale can’t fade or wipe off with scrubbing or exposure to high temperatures
  • Ideal for a variety of lab purposes, including:
    • Preparing solutions and decanting supernatant fluids
    • Holding and measuring waste fluids prior to disposal
    • Measuring solutions for simple chemical reactions
  • FDA-certified for contact with all aqueous, alcoholic and fatty foods