missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAMSN1 Antibody (OTI7F5), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73984AF405
This item is not returnable.
View return policy
Description
SAMSN1 Monoclonal specifically detects SAMSN1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| SAMSN1 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SAMSN1 | |
| Recombinant protein fragment corresponding to amino acids 1-250 of human SAMSN1 (NP_071419) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI7F5 | |
| Western Blot, Immunohistochemistry | |
| HACS1gene with homology to KIAA0790 protein10NASH1, Hematopoietic adaptor containing SH3 and SAM domains 1, SAM and SH3 domain containing 1, SAM and SH3 domain containing 2, SAM domain, SH3 domain and nuclear localisation signals, 1, SAM domain, SH3 domain and nuclear localization signals 1, SAM domain, SH3 domain and nuclear localization signals protein 1, SAM domain-containing protein SAMSN-1, SH3D6B, SH3-SAM adaptor protein | |
| Mouse | |
| Affinity Purified | |
| Stem Cell Markers | |
| 64092.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction