missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAMSN1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97172-50ul
This item is not returnable.
View return policy
Description
SAMSN1 Polyclonal antibody specifically detects SAMSN1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SAMSN1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:500-1:2000 | |
| HACS1gene with homology to KIAA0790 protein10NASH1, Hematopoietic adaptor containing SH3 and SAM domains 1, SAM and SH3 domain containing 1, SAM and SH3 domain containing 2, SAM domain, SH3 domain and nuclear localisation signals, 1, SAM domain, SH3 domain and nuclear localization signals 1, SAM domain, SH3 domain and nuclear localization signals protein 1, SAM domain-containing protein SAMSN-1, SH3D6B, SH3-SAM adaptor protein | |
| Produced in rabbits immunized with E. coli-derived Human SAMSN1 fragment. | |
| 50 μg | |
| Stem Cell Markers | |
| 64092 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction